gRNA Design Details
- Target DNA Sequence
- GATCCATACAGGGCCCCATC
- PAM Sequence
- TGG
- Target locus
-
Chr.19: 48757006 - 48757028 on GRCh38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
88.52/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- FGF21
- Entrez Gene ID
-
26291
- Gene Name
- fibroblast growth factor 21
- Gene Aliases
-
-
- Chromosome Location
-
Chr.19: 48755559 - 48758330 on GRCh38
- UniGene ID
- Hs.283015
- Species
-
Human