gRNA Design Details

Target DNA Sequence
GCCAAACGCTGGACATTAGT
PAM Sequence
GGG
Target locus
Chr.3: 41225067 - 41225089 on GRCh38
Strand
Reverse
Application
Gene Knock-Out
Score: 93.4/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
CTNNB1
Entrez Gene ID
1499
Gene Name
catenin beta 1
Gene Aliases
CTNNB, MRD19, armadillo
Chromosome Location
Chr.3: 41199451 - 41240448 on GRCh38
UniGene ID
Hs.476018
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001098209.1 NP_001091679.1 4
NM_001098210.1 NP_001091680.1 4
NM_001904.3 NP_001895.1 4
XM_005264886.2 XP_005264943.1 4
XM_006712983.1 XP_006713046.1 4
XM_006712984.1 XP_006713047.1 4
XM_006712985.1 XP_006713048.1 4
XM_017005738.1 XP_016861227.1 4