Targeted Isoforms | Translated Protein | Targeted Exon(s) | |
---|---|---|---|
RefSeq | NM_001278547.1 | NP_001265476.1 | 3 |
NM_001278548.1 | NP_001265477.1 | 2 | |
NM_001323302.1 | NP_001310231.1 | 5 | |
NM_001323320.1 | NP_001310249.1 | 3 | |
NM_001323321.1 | NP_001310250.1 | 3 | |
NM_001323322.1 | NP_001310251.1 | 5 | |
NM_001323323.1 | NP_001310252.1 | 4 | |
NM_001323324.1 | NP_001310253.1 | 3 | |
NM_001323325.1 | NP_001310254.1 | 4 | |
NM_001323326.1 | NP_001310255.1 | 4 | |
NM_001323327.1 | NP_001310256.1 | 4 | |
NM_001323328.1 | NP_001310257.1 | 4 | |
NM_001323329.1 | NP_001310258.1 | 3 | |
NM_001323330.1 | NP_001310259.1 | 4 | |
NM_001323331.1 | NP_001310260.1 | 4 | |
NM_139046.3 | NP_620634.1 | 5 | |
NM_139049.3 | NP_620637.1 | 5 | |
NR_136583.1 | 3 | ||
NR_136584.1 | 3 | ||
NR_136585.1 | 3 |