gRNA Design Details
- Target DNA Sequence
- ATGTGAAGCTTGGATGGCCA
- PAM Sequence
- AGG
- Target locus
-
Chr.5: 90619099 - 90619121 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
87.07/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- ADGRV1
- Entrez Gene ID
-
84059
- Gene Name
- adhesion G protein-coupled receptor V1
- Gene Aliases
-
FEB4, GPR98, MASS1, USH2B, USH2C, VLGR1, VLGR1b
- Chromosome Location
-
Chr.5: 90558681 - 91164226 on GRCh38
- UniGene ID
- Hs.591777
- Species
-
Human