gRNA Design Details

Target DNA Sequence
GCTGCAGCTTTTATACCTGC
PAM Sequence
CGG
Target locus
Chr.5: 90617843 - 90617865 on GRCh38
Strand
Forward
Application
Gene Knockout
Score: 87.81/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
ADGRV1
Entrez Gene ID
84059
Gene Name
adhesion G protein-coupled receptor V1
Gene Aliases
FEB4, GPR98, MASS1, USH2B, USH2C, VLGR1, VLGR1b
Chromosome Location
Chr.5: 90558681 - 91164226 on GRCh38
UniGene ID
Hs.591777
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq XM_017009974.1 XP_016865463.1 3