gRNA Design Details
- Target DNA Sequence
- TACCCCGGATTGTCAGTGAA
- PAM Sequence
- AGG
- Target locus
-
Chr.6: 83523542 - 83523564 on GRCh38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
90.62/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- PRSS35
- Entrez Gene ID
-
167681
- Gene Name
- protease, serine 35
- Gene Aliases
-
C6orf158, dJ223E3.1
- Chromosome Location
-
Chr.6: 83512475 - 83525704 on GRCh38
- UniGene ID
- Hs.98381
- Species
-
Human