gRNA Design Details

Target DNA Sequence
ATAAACGGTATACCCATGGC
PAM Sequence
CGG
Target locus
Chr.6: 22292622 - 22292644 on GRCh38
Strand
Reverse
Application
Gene Knockout
Score: 97/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
PRL
Entrez Gene ID
5617
Gene Name
prolactin
Gene Aliases
GHA1
Chromosome Location
Chr.6: 22287244 - 22302897 on GRCh38
UniGene ID
Hs.1905
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_000948.5 NP_000939.1 3
NM_001163558.2 NP_001157030.1 4
XM_011514753.2 XP_011513055.1 4
XM_011514754.2 XP_011513056.1 5