gRNA Design Details
- Target DNA Sequence
- TATCTTGGAATTCATCAGTG
- PAM Sequence
- TGG
- Target locus
-
Chr.8: 16995735 - 16995757 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
84.06/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- FGF20
- Entrez Gene ID
-
26281
- Gene Name
- fibroblast growth factor 20
- Gene Aliases
-
FGF-20, RHDA2
- Chromosome Location
-
Chr.8: 16992825 - 17002165 on GRCh38
- UniGene ID
- Hs.199905
- Species
-
Human