gRNA Design Details
- Target DNA Sequence
- AGTATGCCAATGACCAGGAG
- PAM Sequence
- CGG
- Target locus
-
Chr.1: 11046990 - 11047012 on GRCh38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
91.06/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- MASP2
- Entrez Gene ID
-
10747
- Gene Name
- mannan binding lectin serine peptidase 2
- Gene Aliases
-
MAP19, MASP-2, MASP1P1, sMAP
- Chromosome Location
-
Chr.1: 11026523 - 11047239 on GRCh38
- UniGene ID
- Hs.655645
- Species
-
Human