Targeted Isoforms | Translated Protein | Targeted Exon(s) | |
---|---|---|---|
RefSeq | NM_001288824.2 | NP_001275753.1 | 3 |
NM_001288825.2 | NP_001275754.1 | 4 | |
NM_001288826.2 | NP_001275755.1 | 4 | |
NM_001288827.2 | NP_001275756.1 | 4 | |
NM_001288828.2 | NP_001275757.1 | 4 | |
NM_005260.4 | NP_005251.1 | 2 | |
XM_005271957.4 | XP_005272014.1 | 3 | |
XM_006714585.3 | XP_006714648.1 | 3 | |
XM_011543308.2 | XP_011541610.1 | 3 | |
XM_011543309.1 | XP_011541611.1 | 4 | |
XM_011543310.1 | XP_011541612.1 | 4 | |
XM_011543311.2 | XP_011541613.1 | 2 |