gRNA Design Details
- Target DNA Sequence
- GTCCTCTTGATTCTGATAGA
- PAM Sequence
- AGG
- Target locus
-
Chr.3: 125029409 - 125029431 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
86.89/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- HEG1
- Entrez Gene ID
-
57493
- Gene Name
- heart development protein with EGF like domains 1
- Gene Aliases
-
HEG, MST112, MSTP112
- Chromosome Location
-
Chr.3: 124965710 - 125055958 on GRCh38
- UniGene ID
- Hs.477420
- Species
-
Human