gRNA Design Details
- Target DNA Sequence
- TTGGCCAGAAAGTAACACTG
- PAM Sequence
- AGG
- Target locus
-
Chr.3: 125029450 - 125029472 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
86.37/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- HEG1
- Entrez Gene ID
-
57493
- Gene Name
- heart development protein with EGF like domains 1
- Gene Aliases
-
HEG, MST112, MSTP112
- Chromosome Location
-
Chr.3: 124965710 - 125055958 on GRCh38
- UniGene ID
- Hs.477420
- Species
-
Human