gRNA Design Details
- Target DNA Sequence
- TTCACTTACGCCTCTCAAGG
- PAM Sequence
- AGG
- Target locus
-
Chr.19: 4668605 - 4668627 on GRCh38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
89.08/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- MYDGF
- Entrez Gene ID
-
56005
- Gene Name
- myeloid derived growth factor
- Gene Aliases
-
C19orf10, EUROIMAGE1875335, IL25, IL27, IL27w, R33729_1, SF20
- Chromosome Location
-
Chr.19: 4657545 - 4670403 on GRCh38
- UniGene ID
- Hs.465645
- Species
-
Human