gRNA Design Details

Target DNA Sequence
GATACGACCGATGAGGACAC
PAM Sequence
GGG
Target locus
Chr.2: 180589553 - 180589575 on GRCm38
Strand
Forward
Application
Gene Knock-Out
Score: 89.66/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
Ogfr
Entrez Gene ID
72075
Gene Name
opioid growth factor receptor
Gene Aliases
2010013E17Rik
Chromosome Location
Chr.2: 180589407 - 180595837 on GRCm38
UniGene ID
Mm.250418
Species
Mouse

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_031373.3 NP_113550.3 1