gRNA Design Details
- Target DNA Sequence
- GAATTCGCCCCTGTGGAAGA
- PAM Sequence
- TGG
- Target locus
-
Chr.1: 115286397 - 115286419 on GRCh38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
90.36/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- NGF
- Entrez Gene ID
-
4803
- Gene Name
- nerve growth factor
- Gene Aliases
-
Beta-NGF, HSAN5, NGFB
- Chromosome Location
-
Chr.1: 115285915 - 115338253 on GRCh38
- UniGene ID
- Hs.2561
- Species
-
Human