gRNA Design Details
- Target DNA Sequence
- GGTAACAGACACCTCTGGCT
- PAM Sequence
- GGG
- Application
- Gene Knock-Out
Score:
87.62/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- ELK1
- Entrez Gene ID
-
2002
- Gene Name
- ELK1, ETS transcription factor
- Gene Aliases
-
-
- Chromosome Location
-
Chr.X: 47635520 - 47650604 on GRCh38
- UniGene ID
- Hs.181128
- Species
-
Human