gRNA Design Details
- Target DNA Sequence
- CTTTAACTTGTCCGCGATGG
- PAM Sequence
- TGG
- Target locus
-
Chr.8: 95571139 - 95571161 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
86.05/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Prss54
- Entrez Gene ID
-
70993
- Gene Name
- protease, serine 54
- Gene Aliases
-
4931432M23Rik, Klkbl4
- Chromosome Location
-
Chr.8: 95559067 - 95576330 on GRCm38
- UniGene ID
- Mm.486561
- Species
-
Mouse