gRNA Design Details
- Target DNA Sequence
- GCCACGAACATCAGGAATTA
- PAM Sequence
- CGG
- Target locus
-
Chr.10: 133127285 - 133127307 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
92.56/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- ADGRA1
- Entrez Gene ID
-
84435
- Gene Name
- adhesion G protein-coupled receptor A1
- Gene Aliases
-
GPR123
- Chromosome Location
-
Chr.10: 133070929 - 133131675 on GRCh38
- UniGene ID
- Hs.435183
- Species
-
Human