Targeted Isoforms | Translated Protein | Targeted Exon(s) | |
---|---|---|---|
RefSeq | NM_001007225.1 | NP_001007226.1 | 1 |
NM_001291869.1 | NP_001278798.1 | 1 | |
NM_006548.4 | NP_006539.3 | 1 | |
XM_017005557.1 | XP_016861046.1 | 1 | |
XM_017005558.1 | XP_016861047.1 | 1 | |
XM_017005559.1 | XP_016861048.1 | 1 | |
XM_017005561.1 | XP_016861050.1 | 1 | |
XM_017005562.1 | XP_016861051.1 | 1 | |
XM_017005563.1 | XP_016861052.1 | 1 | |
XM_017005564.1 | XP_016861053.1 | 1 |