gRNA Design Details
- Target DNA Sequence
- TGCAGGATGCGGTACCGTTT
- PAM Sequence
- GGG
- Target locus
-
Chr.13: 21671950 - 21671972 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
94.55/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- FGF9
- Entrez Gene ID
-
2254
- Gene Name
- fibroblast growth factor 9
- Gene Aliases
-
FGF-9, GAF, HBFG-9, HBGF-9, SYNS3
- Chromosome Location
-
Chr.13: 21671076 - 21704501 on GRCh38
- UniGene ID
- Hs.111
- Species
-
Human