gRNA Design Details

Target DNA Sequence
AACATTGACGGAGAAGTAGG
PAM Sequence
TGG
Target locus
Chr.8: 38429686 - 38429708 on GRCh38
Strand
Forward
Application
Gene Knock-Out
Score: 88.9/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
FGFR1
Entrez Gene ID
2260
Gene Name
fibroblast growth factor receptor 1
Gene Aliases
BFGFR, CD331, CEK, ECCL, FGFBR, FGFR-1, FLG, FLT-2, FLT2, HBGFR, HH2, HRTFDS, KAL2, N-SAM, OGD, bFGF-R-1
Chromosome Location
Chr.8: 38411138 - 38468834 on GRCh38
UniGene ID
Hs.264887
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001174063.1 NP_001167534.1 3
NM_001174064.1 NP_001167535.1 4
NM_001174065.1 NP_001167536.1 3
NM_001174067.1 NP_001167538.1 4
NM_015850.3 NP_056934.2 3
NM_023110.2 NP_075598.2 3
XM_006716303.2 XP_006716366.1 3
XM_006716304.1 XP_006716367.1 3
XM_006716306.2 XP_006716369.1 3
XM_006716307.1 XP_006716370.1 3
XM_006716309.3 XP_006716372.1 3
XM_011544443.2 XP_011542745.1 3
XM_011544444.1 XP_011542746.1 3
XM_011544445.2 XP_011542747.1 3
XM_011544446.2 XP_011542748.1 3
XM_011544447.2 XP_011542749.1 3
XM_011544452.2 XP_011542754.1 3
XM_017013219.1 XP_016868708.1 3
XM_017013220.1 XP_016868709.1 3
XM_017013221.1 XP_016868710.1 4
XM_017013222.1 XP_016868711.1 3
XM_017013223.1 XP_016868712.1 3
XM_017013224.1 XP_016868713.1 3
XM_017013225.1 XP_016868714.1 3
XM_017013230.1 XP_016868719.1 1
XM_017013231.1 XP_016868720.1 3