gRNA Design Details
- Target DNA Sequence
- AACATTGACGGAGAAGTAGG
- PAM Sequence
- TGG
- Target locus
-
Chr.8: 38429686 - 38429708 on GRCh38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
88.9/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- FGFR1
- Entrez Gene ID
-
2260
- Gene Name
- fibroblast growth factor receptor 1
- Gene Aliases
-
BFGFR, CD331, CEK, ECCL, FGFBR, FGFR-1, FLG, FLT-2, FLT2, HBGFR, HH2, HRTFDS, KAL2, N-SAM, OGD, bFGF-R-1
- Chromosome Location
-
Chr.8: 38411138 - 38468834 on GRCh38
- UniGene ID
- Hs.264887
- Species
-
Human