gRNA Design Details
- Target DNA Sequence
- TGACGGCCCTCAGCAGGTTT
- PAM Sequence
- TGG
- Target locus
-
Chr.3: 159990273 - 159990295 on GRCh38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
86.72/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- IL12A
- Entrez Gene ID
-
3592
- Gene Name
- interleukin 12A
- Gene Aliases
-
CLMF, IL-12A, NFSK, NKSF1, P35
- Chromosome Location
-
Chr.3: 159988836 - 159996019 on GRCh38
- UniGene ID
- Hs.673
- Species
-
Human