gRNA Design Details

Target DNA Sequence
TCTCAGCGTGGCATACACAT
PAM Sequence
TGG
Target locus
Chr.1: 6571046 - 6571068 on GRCh38
Strand
Reverse
Application
Gene Knock-Out
Score: 90.86/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
TAS1R1
Entrez Gene ID
80835
Gene Name
taste 1 receptor member 1
Gene Aliases
GM148, GPR70, T1R1, TR1
Chromosome Location
Chr.1: 6555278 - 6579757 on GRCh38
UniGene ID
Hs.124574
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_138697.3 NP_619642.2 2
NM_177540.2 NP_803884.1 2
XM_011542203.2 XP_011540505.1 2
XM_011542206.2 XP_011540508.1 1
XM_017002402.1 XP_016857891.1 1
XM_017002403.1 XP_016857892.1 1