gRNA Design Details
- Target DNA Sequence
- GCCCATGAGCTCCACAGTCA
- PAM Sequence
- AGG
- Target locus
-
Chr.11: 64235873 - 64235895 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
84.7/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- VEGFB
- Entrez Gene ID
-
7423
- Gene Name
- vascular endothelial growth factor B
- Gene Aliases
-
VEGFL, VRF
- Chromosome Location
-
Chr.11: 64234584 - 64239264 on GRCh38
- UniGene ID
- Hs.732095
- Species
-
Human