gRNA Design Details
- Target DNA Sequence
- CTCTCTTCCAAATTTGAGCA
- PAM Sequence
- TGG
- Target locus
-
Chr.9: 17786480 - 17786502 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
83.17/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- SH3GL2
- Entrez Gene ID
-
6456
- Gene Name
- SH3 domain containing GRB2 like 2, endophilin A1
- Gene Aliases
-
CNSA2, EEN-B1, SH3D2A, SH3P4
- Chromosome Location
-
Chr.9: 17578954 - 17797129 on GRCh38
- UniGene ID
- Hs.743740
- Species
-
Human