gRNA Design Details
- Target DNA Sequence
- GAATTTCGGTGGCGGCAACA
- PAM Sequence
- CGG
- Target locus
-
Chr.15: 88819954 - 88819976 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
93.45/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Creld2
- Entrez Gene ID
-
76737
- Gene Name
- cysteine-rich with EGF-like domains 2
- Gene Aliases
-
5730592L21Rik, C85758
- Chromosome Location
-
Chr.15: 88819646 - 88826681 on GRCm38
- UniGene ID
- Mm.292567
- Species
-
Mouse