gRNA Design Details
- Target DNA Sequence
- TCTAGACTCAGATGAGAGTG
- PAM Sequence
- AGG
- Target locus
-
Chr.7: 99403418 - 99403440 on GRCh38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
70.33/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- PDAP1
- Entrez Gene ID
-
11333
- Gene Name
- PDGFA associated protein 1
- Gene Aliases
-
HASPP28, PAP, PAP1
- Chromosome Location
-
Chr.7: 99394673 - 99408682 on GRCh38
- UniGene ID
- Hs.632296
- Species
-
Human