gRNA Design Details
- Target DNA Sequence
- GACGTGGATGAGTGCTCAGA
- PAM Sequence
- GGG
- Target locus
-
Chr.22: 43339205 - 43339227 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
89.01/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- SCUBE1
- Entrez Gene ID
-
80274
- Gene Name
- signal peptide, CUB domain and EGF like domain containing 1
- Gene Aliases
-
-
- Chromosome Location
-
Chr.22: 43203223 - 43343388 on GRCh38
- UniGene ID
- Hs.133995
- Species
-
Human