gRNA Design Details
- Target DNA Sequence
- TCATCGAGGTCCCATCAGGT
- PAM Sequence
- GGG
- Application
- Gene Knock-Out
Score:
90.62/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- IKBKG
- Entrez Gene ID
-
8517
- Gene Name
- inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase gamma
- Gene Aliases
-
AMCBX1, FIP-3, FIP3, Fip3p, IKK-gamma, IKKAP1, IKKG, IMD33, IP, IP1, IP2, IPD2, NEMO, ZC2HC9
- Chromosome Location
-
Chr.X: 154542240 - 154565046 on GRCh38
- UniGene ID
- Hs.43505
- Species
-
Human