gRNA Design Details
- Target DNA Sequence
- ATGGTCCATATCTGCCCACT
- PAM Sequence
- GGG
- Target locus
-
Chr.3: 100630754 - 100630776 on GRCh38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
88.78/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- ADGRG7
- Entrez Gene ID
-
84873
- Gene Name
- adhesion G protein-coupled receptor G7
- Gene Aliases
-
GPR128
- Chromosome Location
-
Chr.3: 100609589 - 100696174 on GRCh38
- UniGene ID
- Hs.334511
- Species
-
Human