gRNA Design Details

Target DNA Sequence
AGAAGACTATCCTGAGCCCG
PAM Sequence
AGG
Target locus
Chr.1: 218346863 - 218346885 on GRCh38
Strand
Forward
Application
Gene Knock-Out
Score: 90.79/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
TGFB2
Entrez Gene ID
7042
Gene Name
transforming growth factor beta 2
Gene Aliases
LDS4, TGF-beta2
Chromosome Location
Chr.1: 218345334 - 218444619 on GRCh38
UniGene ID
Hs.133379
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001135599.2 NP_001129071.1 1
NM_003238.3 NP_003229.1 1