gRNA Design Details
- Target DNA Sequence
- TCCCGGGATTTTATGGTACC
- PAM Sequence
- GGG
- Target locus
-
Chr.6: 31972388 - 31972410 on GRCh38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
95.36/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- STK19
- Entrez Gene ID
-
8859
- Gene Name
- serine/threonine kinase 19
- Gene Aliases
-
D6S60, D6S60E, G11, HLA-RP1, RP1
- Chromosome Location
-
Chr.6: 31971175 - 31981446 on GRCh38
- UniGene ID
- Hs.654371
- Species
-
Human