gRNA Design Details
- Target DNA Sequence
- TCCCCGGTACCATAAAATCC
- PAM Sequence
- CGG
- Target locus
-
Chr.6: 31972386 - 31972408 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
92.39/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- STK19
- Entrez Gene ID
-
8859
- Gene Name
- serine/threonine kinase 19
- Gene Aliases
-
D6S60, D6S60E, G11, HLA-RP1, RP1
- Chromosome Location
-
Chr.6: 31971175 - 31981446 on GRCh38
- UniGene ID
- Hs.654371
- Species
-
Human