gRNA Design Details

Target DNA Sequence
GCGGCATCTAGTAGAAAAGG
PAM Sequence
GGG
Target locus
Chr.2: 218134889 - 218134911 on GRCh38
Strand
Reverse
Application
Gene Knockout
Score: 93.75/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
CXCR2
Entrez Gene ID
3579
Gene Name
C-X-C motif chemokine receptor 2
Gene Aliases
CD182, CDw128b, CMKAR2, IL8R2, IL8RA, IL8RB
Chromosome Location
Chr.2: 218125290 - 218137253 on GRCh38
UniGene ID
Hs.846
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001168298.1 NP_001161770.1 4
NM_001557.3 NP_001548.1 3
XM_005246530.3 XP_005246587.1 4
XM_017003990.1 XP_016859479.1 4
XM_017003991.1 XP_016859480.1 2
XM_017003992.1 XP_016859481.1 2