gRNA Design Details
- Target DNA Sequence
- GTGGTGTCTGCTATATCCGT
- PAM Sequence
- GGG
- Target locus
-
Chr.3: 32953429 - 32953451 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
91.48/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- CCR4
- Entrez Gene ID
-
1233
- Gene Name
- C-C motif chemokine receptor 4
- Gene Aliases
-
CC-CKR-4, CD194, CKR4, CMKBR4, ChemR13, HGCN:14099, K5-5
- Chromosome Location
-
Chr.3: 32951555 - 32955312 on GRCh38
- UniGene ID
- Hs.184926
- Species
-
Human