gRNA Design Details

Target DNA Sequence
AGAACTTGCCAGCGAAGTCC
PAM Sequence
GGG
Target locus
Chr.19: 40605099 - 40605121 on GRCh38
Strand
Reverse
Application
Gene Knock-Out
Score: 85.28/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
LTBP4
Entrez Gene ID
8425
Gene Name
latent transforming growth factor beta binding protein 4
Gene Aliases
ARCL1C, LTBP-4, LTBP4L, LTBP4S
Chromosome Location
Chr.19: 40592888 - 40629820 on GRCh38
UniGene ID
Hs.466766
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001042544.1 NP_001036009.1 5
NM_001042545.1 NP_001036010.1 2
NM_003573.2 NP_003564.2 5
XM_011527376.2 XP_011525678.1 5
XM_011527377.2 XP_011525679.1 5
XM_011527378.2 XP_011525680.1 5
XM_011527379.1 XP_011525681.1 2
XM_011527380.2 XP_011525682.1 5
XM_011527381.2 XP_011525683.1 5
XM_011527382.2 XP_011525684.1 5
XM_011527383.2 XP_011525685.1 5
XM_011527384.2 XP_011525686.1 5
XM_011527385.2 XP_011525687.1 5
XM_011527386.2 XP_011525688.1 5
XM_011527387.1 XP_011525689.1 5
XM_017027352.1 XP_016882841.1 5
XM_017027353.1 XP_016882842.1 5
XM_017027354.1 XP_016882843.1 5