gRNA Design Details
- Target DNA Sequence
- TTCCGACACTGTAACCCCAA
- PAM Sequence
- TGG
- Target locus
-
Chr.2: 208437768 - 208437790 on GRCh38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
92.25/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- PTH2R
- Entrez Gene ID
-
5746
- Gene Name
- parathyroid hormone 2 receptor
- Gene Aliases
-
PTHR2
- Chromosome Location
-
Chr.2: 208359845 - 208494506 on GRCh38
- UniGene ID
- Hs.570296
- Species
-
Human