gRNA Design Details

Target DNA Sequence
TGCTGTAGAGTTGGTACTCG
PAM Sequence
CGG
Target locus
Chr.8: 22046197 - 22046219 on GRCh38
Strand
Reverse
Application
Gene Knock-Out
Score: 97.82/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
FGF17
Entrez Gene ID
8822
Gene Name
fibroblast growth factor 17
Gene Aliases
FGF-13, FGF-17, HH20
Chromosome Location
Chr.8: 22039708 - 22048809 on GRCh38
UniGene ID
Hs.248192
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001304478.1 NP_001291407.1 3
NM_003867.3 NP_003858.1 3
XM_005273675.1 XP_005273732.1 1
XM_011544683.1 XP_011542985.1 3
XM_011544684.2 XP_011542986.1 4
XM_011544685.1 XP_011542987.1 3