gRNA Design Details
- Target DNA Sequence
- GGCAATAAACCGAGTGGCGA
- PAM Sequence
- TGG
- Target locus
-
Chr.2: 232521376 - 232521398 on GRCh38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
90.4/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- PRSS56
- Entrez Gene ID
-
646960
- Gene Name
- protease, serine 56
- Gene Aliases
-
MCOP6
- Chromosome Location
-
Chr.2: 232520463 - 232525716 on GRCh38
- UniGene ID
- Hs.570310
- Species
-
Human