gRNA Design Details
- Target DNA Sequence
- GAGGTACTTCAGCAGCGTGG
- PAM Sequence
- AGG
- Target locus
-
Chr.2: 26183013 - 26183035 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
89.43/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- GAREM2
- Entrez Gene ID
-
150946
- Gene Name
- GRB2 associated regulator of MAPK1 subtype 2
- Gene Aliases
-
FAM59B, GAREML
- Chromosome Location
-
Chr.2: 26173091 - 26202405 on GRCh38
- UniGene ID
- Hs.187912
- Species
-
Human