gRNA Design Details
- Target DNA Sequence
- TGTAGGAGTGATTGTCCTCA
- PAM Sequence
- AGG
- Target locus
-
Chr.20: 62809634 - 62809656 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
87.21/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- OGFR
- Entrez Gene ID
-
11054
- Gene Name
- opioid growth factor receptor
- Gene Aliases
-
-
- Chromosome Location
-
Chr.20: 62804825 - 62814000 on GRCh38
- UniGene ID
- Hs.67896
- Species
-
Human