gRNA Design Details
- Target DNA Sequence
- GGGTGAGCTATTCGACTACC
- PAM Sequence
- TGG
- Target locus
-
Chr.19: 55289507 - 55289529 on GRCh38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
90.76/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- BRSK1
- Entrez Gene ID
-
84446
- Gene Name
- BR serine/threonine kinase 1
- Gene Aliases
-
hSAD1
- Chromosome Location
-
Chr.19: 55284101 - 55312562 on GRCh38
- UniGene ID
- Hs.182081
- Species
-
Human