gRNA Design Details

Target DNA Sequence
ACAGAAGGACACCATCCCGG
PAM Sequence
AGG
Target locus
Chr.19: 6772915 - 6772937 on GRCh38
Strand
Reverse
Application
Gene Knockout
Score: 87.02/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
VAV1
Entrez Gene ID
7409
Gene Name
vav guanine nucleotide exchange factor 1
Gene Aliases
VAV
Chromosome Location
Chr.19: 6772668 - 6857366 on GRCh38
UniGene ID
Hs.116237
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001258206.1 NP_001245135.1 1
NM_001258207.1 NP_001245136.1 1
NM_005428.3 NP_005419.2 1
XM_005259642.1 XP_005259699.1 1