Targeted Isoforms | Translated Protein | Targeted Exon(s) | |
---|---|---|---|
RefSeq | NM_000142.4 | NP_000133.1 | 3 |
NM_001163213.1 | NP_001156685.1 | 3 | |
NM_022965.3 | NP_075254.1 | 3 | |
XM_006713868.1 | XP_006713931.1 | 3 | |
XM_006713869.1 | XP_006713932.1 | 3 | |
XM_006713870.1 | XP_006713933.1 | 3 | |
XM_006713871.1 | XP_006713934.1 | 3 | |
XM_006713872.1 | XP_006713935.1 | 3 | |
XM_006713873.1 | XP_006713936.1 | 3 | |
XM_011513420.1 | XP_011511722.1 | 3 | |
XM_011513422.1 | XP_011511724.1 | 3 |