gRNA Design Details
- Target DNA Sequence
- CCGGAACAGACTGTACACCT
- PAM Sequence
- TGG
- Target locus
-
Chr.14: 74611726 - 74611748 on GRCh38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
86.49/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- LTBP2
- Entrez Gene ID
-
4053
- Gene Name
- latent transforming growth factor beta binding protein 2
- Gene Aliases
-
C14orf141, GLC3D, LTBP3, MSPKA, MSTP031, WMS3
- Chromosome Location
-
Chr.14: 74498183 - 74612331 on GRCh38
- UniGene ID
- Hs.512776
- Species
-
Human