gRNA Design Details

Target DNA Sequence
AAGAGGTTGGGGCCGAATAA
PAM Sequence
CGG
Target locus
Chr.12: 6556001 - 6556023 on GRCh38
Strand
Forward
Application
Gene Knockout
Score: 88.85/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
IFFO1
Entrez Gene ID
25900
Gene Name
intermediate filament family orphan 1
Gene Aliases
HOM-TES-103, IFFO
Chromosome Location
Chr.12: 6539528 - 6556102 on GRCh38
UniGene ID
Hs.15243
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001039670.2 NP_001034759.1 1
NM_001193457.1 NP_001180386.1 1
NM_080730.4 NP_542768.2 1
NR_036467.1 1
XM_006718975.3 XP_006719038.1 1
XM_006718976.3 XP_006719039.1 1
XM_006718977.3 XP_006719040.1 1
XM_011520947.2 XP_011519249.1 1
XM_011520948.2 XP_011519250.1 1
XM_011520949.2 XP_011519251.1 1
XM_017019169.1 XP_016874658.1 1
XM_017019171.1 XP_016874660.1 1