Targeted Isoforms | Translated Protein | Targeted Exon(s) | |
---|---|---|---|
RefSeq | NM_001039670.2 | NP_001034759.1 | 1 |
NM_001193457.1 | NP_001180386.1 | 1 | |
NM_080730.4 | NP_542768.2 | 1 | |
NR_036467.1 | 1 | ||
XM_006718975.3 | XP_006719038.1 | 1 | |
XM_006718976.3 | XP_006719039.1 | 1 | |
XM_006718977.3 | XP_006719040.1 | 1 | |
XM_011520947.2 | XP_011519249.1 | 1 | |
XM_011520948.2 | XP_011519250.1 | 1 | |
XM_011520949.2 | XP_011519251.1 | 1 | |
XM_017019169.1 | XP_016874658.1 | 1 | |
XM_017019170.1 | XP_016874659.1 | 2 | |
XM_017019171.1 | XP_016874660.1 | 1 |