gRNA Design Details

Target DNA Sequence
GTACTTGGCAATGAACAGTG
PAM Sequence
AGG
Application
Gene Knock-Out
Score: 90.69/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
FGF13
Entrez Gene ID
2258
Gene Name
fibroblast growth factor 13
Gene Aliases
FGF-13, FGF2, FHF-2, FHF2
Chromosome Location
Chr.X: 138631573 - 139222889 on GRCh38
UniGene ID
Hs.6540
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001139498.1 NP_001132970.1 4
NM_001139500.1 NP_001132972.1 5
NM_001139501.1 NP_001132973.1 6
NM_001139502.1 NP_001132974.1 4
NM_004114.3 NP_004105.1 3
NM_033642.2 NP_378668.1 3
XM_005262399.1 XP_005262456.1 5