gRNA Design Details
- Target DNA Sequence
- ACCGTACCACCGAAGATGCA
- PAM Sequence
- GGG
- Target locus
-
Chr.6: 131950507 - 131950529 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
95.91/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- CTGF
- Entrez Gene ID
-
1490
- Gene Name
- connective tissue growth factor
- Gene Aliases
-
CCN2, HCS24, IGFBP8, NOV2
- Chromosome Location
-
Chr.6: 131948176 - 131951378 on GRCh38
- UniGene ID
- Hs.410037
- Species
-
Human